EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Acute toxicity of 2,4,6-trinitrotoluene, 2,4-dinitrotoluene, and 2,6-dinitrotoluene in the adult bullfrog (Lithobates catesbeiana).
PMID 18551238
Abstract
2,4,6-Trinitrotoluene (TNT) is one of the most prevalent high explosives in the environment. 2,4-Dinitrotoluene (2,4-DNT) and 2,6-dinitrotoluene (2,6-DNT) are the most common isoforms of dinitrotoluene. The goal of this study was to determine the acute toxic effects of TNT, 2,4-DNT, and 2,6-DNT in adult male bullfrogs. The LD(50) for TNT was 1,060 mg/kg BW while the LD(50 )for 2,4-DNT and 2,6-DNT was 1,098 mg/kg BW. All three compounds elicited similar symptoms of toxicity including changes of skin color, body weight, development of seizures, liver and kidney necrosis, and lung cyanosis. Relative organ weights did not show significant change.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D200603
2,6-Dinitrotoluene, 98%
C7H6N2O4