EMAIL THIS PAGE TO A FRIEND
Journal of combinatorial chemistry
Solid-phase synthesis of solanesol.
PMID 18558751
Abstract
An efficient and convergent solid-phase strategy for the total synthesis of All- E solanesol is described. This method features avoidance of iterative and difficult purifications comparing with solution-phase synthesis and is suitable for the preparation of other oligoprenols.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8754
Solanesol from tobacco, ≥90% (HPLC)
C45H74O