EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Post-translational enzyme modification by the phosphopantetheinyl transferase is required for lysine and penicillin biosynthesis but not for roquefortine or fatty acid formation in Penicillium chrysogenum.
PMID 18558918
Abstract
NRPSs (non-ribosomal peptide synthetases) and PKSs (polyketide synthases) require post-translational phosphopantetheinylation to become active. This reaction is catalysed by a PPTase (4'-phosphopantetheinyl transferase). The ppt gene of Penicillium chrysogenum, encoding a protein that shares 50% similarity with the stand-alone large PPTases, has been cloned. This gene is present as a single copy in the genome of the wild-type and high-penicillin-producing strains (containing multiple copies of the penicillin gene cluster). Amplification of the ppt gene produced increases in isopenicillin N and benzylpenicillin biosynthesis. A PPTase-defective mutant (Wis54-PPT(-)) was obtained. It required lysine and lacked pigment and penicillin production, but it still synthesized normal levels of roquefortine. The biosynthesis of roquefortine does not appear to involve PPTase-mediated modification of the synthesizing enzymes. The PPT(-) mutant did not require fatty acids, which indicates that activation of the fatty acid synthase is performed by a different PPTase. Complementation of Wis54-PPT(-) with the ppt gene restored lysine biosynthesis, pigmentation and penicillin production, which demonstrates the wide range of processes controlled by this gene.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0406 Roquefortine C, ≥98% (HPLC), from Penicillium roqueforti
C22H23N5O2