EMAIL THIS PAGE TO A FRIEND
Chirality
Enantiomer self-disproportionation of chiral compounds on achiral ordered mesoporous silica M41S and regular silica gel as a stationary phase.
PMID 18561251
Abstract
Chromatographic behavior of nonracemic mixtures, viz., mandelic acid and stilbene oxide as analytes has been studied in detailed by enantiomer self-disproportionation on achiral ordered mesoporous material M41S and regular silica gel as stationary phases. Enantiomer self-disproportionation gave enhanced separation of analytes. The extent and magnitude of enantiomer self-disproportionation is dependent on the optical purity of the starting non-racemic molecules, presence of intermolecular hydrogen bonding/pi-pi interactions and the nature of eluents used. The present study and previous literature data suggest that percentage ee of a nonracemic mixture needs to be determined before any chromatographic purification is taken up as enantiomer self-disproportionation phenomenon could occur during purification. The data show that enantiomer self-disproportionation of nonracemic mixtures can be harnessed for its enantioenrichment on inexpensive achiral stationary phases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
308323
cis-Stilbene oxide, 97%
C14H12O
S4921
trans-Stilbene oxide, 98%
C14H12O