EMAIL THIS PAGE TO A FRIEND
Archives of pharmacal research
Synthesis of bis, tris and tetra(dihydrocaffeoyl)polyamine conjugates as antibacterial agents against VRSA.
PMID 18563350
Abstract
Bis, tris and tetra(dihydrocaffeoyl)polyamine conjugates were synthesized using solid phase synthesis technique. These compounds were screened for antibacterial activity against methicillin-resistant Staphylococcus aureus (MRSA) (11 strains) and vancomycin-resistant S. aureus (VRSA) (4 strains). Bis, tris and tetra(dihydrocaffeoyl)polyamine analogues showed antibacterial activity against VRSA which were better than the reference drugs, vancomycin. Tetra(dihydrocaffeoyl)polyamine conjugate exhibited the highest activity. These compounds showed no cytotoxicity against vero cells.