EMAIL THIS PAGE TO A FRIEND
Tumori
[Pharmacokinetic and pharmacodynamic features of palonosetron].
PMID 18564620
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9571
Dolasetron mesylate hydrate, ≥98% (HPLC), powder
C19H20N2O3 · CH4O3S · xH2O