EMAIL THIS PAGE TO A FRIEND
Anaesthesia and intensive care
Fire-eater's lung.
PMID 18564809
Abstract
Hydrocarbon aspiration from fire-eating can result in severe necrotising pneumonia with complex cyst formation and large pleural effusions. Despite this, there is usually minimal persistent lung injury. We report the progress of two clinical cases, one of whom is the first reported case of citronella oil aspiration.