EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
One year survival and significant reversal of motor deficits in parkinsonian rats transplanted with hESC derived dopaminergic neurons.
PMID 18565328