EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
Lewis base catalyzed 1,3-dithiane addition to carbonyl and imino compounds using 2-trimethylsilyl-1,3-dithiane.
PMID 18567043
Abstract
Lewis base-catalyzed 1,3-dithiane addition to electrophiles such as carbonyl compounds and N-substituted aldimines with 2-trimethylsilyl-1,3-dithiane (TMS-dithiane) is described. By the activation of the carbon-silicon bond in the presence of a Lewis base catalyst such as tetrabutylammonium phenoxide (PhONnBu(4)), a 1,3-dithiane addition reaction proceeded smoothly to afford the corresponding adducts in good to high yields under mild conditions. This synthesis is also applied to the reactions of ketones having alpha-protons, and of N-substituted aldimines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
157872
1,3-Dithiane, 97%
C4H8S2
220817
2-(Trimethylsilyl)-1,3-dithiane, ≥99%
C7H16S2Si