EMAIL THIS PAGE TO A FRIEND
Headache
The possible treatment of migraine with carteolol hydrochloride.
PMID 18567128
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000193 Carteolol for system suitability, European Pharmacopoeia (EP) Reference Standard
C16H24N2O3 · HCl