EMAIL THIS PAGE TO A FRIEND
Chirality
Enantiomer separation of triazole fungicides by high-performance liquid chromatography.
PMID 18570307
Abstract
Enantiomer separation is one of the most important prerequisites for the investigation of environmental enantioselective behaviors for chiral pesticides. In the present study, the enantiomer separation of 7 triazole fungicides, i.e., hexaconazole (1), triadimefon (2), tebuconazole (3), diniconazole (4), flutriafol (5), propiconazole (6), and difenoconazole (7), were evaluated using normal phase high-performance liquid chromatography. Chrialcel OD column and Chrialcel OJ column were used. The influence of column temperature was studied for the optimization of the resolution as well as the type and percentage of organic modifier in the mobile phase. The retention factors for the enantiomers of all investigated compounds decreased as the temperature increased. The natural logarithms of the selectivity factors (lnalpha) of hexaconazole (1), tebuconazole (3), flutriafol (5), propiconazole (6) and difenoconazole (7) depended linearly on the inverse of temperature (1/T) while the corresponding values for triadimefon (2) and diniconazole (4) kept unchanged in the studied temperature range 10-35 degrees C. Van't Hoff plots afforded thermodynamic parameters, such as the apparent change in enthalpy DeltaH degrees , the apparent change in entropy DeltaS degrees and the apparent change in DeltaDeltaH degrees and DeltaDeltaS degrees . The thermodynamic parameters (DeltaH degrees , DeltaS degrees , DeltaDeltaH degrees and DeltaDeltaS degrees ) were calculated in order to provide an understanding of the thermosynamic driving forces for enantioseparation. The established method shows perspective to be used for preparing micro-scale amount of pure enantiomers of the chiral triazoles studied.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36531
Difenoconazol, PESTANAL®, analytical standard
C19H17Cl2N3O3
34348
Hexaconazol, PESTANAL®, analytical standard
C14H17Cl2N3O