EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Hypaphorine, an indole alkaloid from Erythrina velutina, induced sleep on normal mice.
PMID 18571406
Abstract
An indole alkaloid (hypaphorine (1)) was isolated from Brazilian medicinal plant, Erythrina velutina (Leguminosae). This compound was investigated for sleep promoting effects in mice, and the results showed that it significantly increased non-rapid eye movement (NREM) sleep time during the first hour after its administration. The NREM sleep time was enhanced by 33% in the experimental mice when compared to that of the controls. This study therefore confirmed its sleep promoting property.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3630
Diphenhydramine hydrochloride, ≥98% (HPLC)
C17H21NO · HCl