EMAIL THIS PAGE TO A FRIEND
Journal of environmental sciences (China)
Polycyclic aromatic hydrocarbon biodegradation and extracellular enzyme secretion in agitated and stationary cultures of Phanerochaete chrysosporium.
PMID 18572528
Abstract
The extracellular enzyme secretion and biodegradation of polycyclic aromatic hydrocarbons (PAHs) were studied in agitated and shallow stationary liquid cultures of Phanerochaete chrysosporium. Veratryl alcohol and Tween80 were added to cultures as lignin peroxidase (LiP) and manganese peroxidase (MnP) inducer, respectively. Shallow stationary cultures were suitable for the production of enzyme, whereas agitated cultures enhanced overall biodegradation by facilitating interphase mass transfer of PAHs into aqueous phases. The use of a LiP stimulator, veratryl alcohol, did not increase PAH degradation but significantly enhanced LiP activity. In contrast, Tween80 increased both MnP secretion and PAH degradation in shallow stationary cultures. On the other hand, high PAH degradation was observed in agitated cultures in the absence of detectable LiP and MnP activities. The results suggested that extracellular peroxidase activities are not directly related to the PAH degradation, and the increased solubility rather than enzyme activity may be more important in the promotion of PAH degradation.