EMAIL THIS PAGE TO A FRIEND
Human molecular genetics
A novel mouse model with impaired dynein/dynactin function develops amyotrophic lateral sclerosis (ALS)-like features in motor neurons and improves lifespan in SOD1-ALS mice.
PMID 18579581
Abstract
Amyotrophic lateral sclerosis (ALS) is a fatal neurodegenerative condition characterized by progressive motor neuron degeneration and muscle paralysis. Genetic evidence from man and mouse has indicated that mutations in the dynein/dynactin motor complex are correlated with motor neuron degeneration. In this study, we have generated transgenic mice with neuron-specific expression of Bicaudal D2 N-terminus (BICD2-N) to chronically impair dynein/dynactin function. Motor neurons expressing BICD2-N showed accumulation of dynein and dynactin in the cell body, Golgi fragmentation and several signs of impaired retrograde trafficking: the appearance of giant neurofilament swellings in the proximal axon, reduced retrograde labelling by tracer injected in the muscle and delayed expression of the injury transcription factor ATF3 after axon transection. Despite these abnormalities, BICD2-N mice did not develop signs of motor neuron degeneration and motor abnormalities. Interestingly, the BICD2-N transgene increased lifespan in 'low copy' SOD1-G93A ALS transgenic mice. Our findings indicate that impaired dynein/dynactin function can explain several pathological features observed in ALS patients, but may be beneficial in some forms of ALS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500984
Anti-BICD2 antibody produced in rabbit, affinity isolated antibody