Toxicological sciences : an official journal of the Society of Toxicology
Inhalation dosimetry modeling with decamethylcyclopentasiloxane in rats and humans.
PMID 18583370
Decamethylcyclopentasiloxane (D(5)), a volatile cyclic methyl siloxane (VCMS), is used in industrial and consumer products. Inhalation pharmacokinetics of another VCMS, octamethylcyclotetrasiloxane (D(4)), have been extensively investigated and successfully modeled with a multispecies physiologically based pharmacokinetic (PBPK) model. Here, we develop an inhalation PBPK description for D(5), using the D(4) model structure as a starting point, with the objective of understanding factors that regulate free blood and tissue concentrations of this highly lipophilic vapor after inhalation in rats and humans. Compared with D(4), the more lipophilic D(5) required deep compartments in lung, liver, and plasma to account for slow release from tissues after cessation of exposures. Simulations of the kinetics of a stable D(5) metabolite, HO-D(5), required diffusion-limited uptake in fat, a deep tissue store in lung, and its elimination by fecal excretion and metabolism to linear silanols. The combined D(5)/HO-D(5) model described blood and tissue concentrations of parent D(5) and elimination of total radioactivity in single and repeat exposures in male and female rats at 7 and 160 ppm. In humans, D(5) kinetic data are more sparse and the model structure though much simplified, still required free and bound blood D(5) to simulate exhaled air and blood time courses from 1 h inhalation exposures at 10 ppm in five human volunteers. This multispecies PBPK model for D(5) highlights complications in interpreting kinetic studies where chemical in blood and tissues represents various pools with only a portion free. The ability to simulate free concentrations is essential for dosimetry based risk assessments for these VCMS.
Related Materials
Product #
Molecular Formula
Add to Cart
Octamethylcyclotetrasiloxane, 98%