EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. A
Ozonolysis of geraniol-trans, 6-methyl-5-hepten-2-one, and 6-hydroxy-4-methyl-4-hexenal: kinetics and mechanisms.
PMID 18588276
Abstract
A combined density functional theory and transition-state theory study of the mechanisms and reaction coefficients of gas-phase ozonolysis of geraniol-trans, 6-methyl-5-hepten-2-one, and 6-hydroxy-4-methyl-4-hexenal is presented. The geometries, energies, and harmonic vibrational frequencies of each stationary point were determined by B3LYP/6-31(d,p), MPW1K/cc-pVDZ, and BH&HLYP/cc-pVDZ methods. According to the calculations, the ozone 6-methyl-5-hepten-2-one reaction is faster than the ozone 6-hydroxy-4-methyl-4-hexenal reaction, but both are slower than the ozone geraniol-trans reaction. By using the BH&HLYP/cc-pVDZ data, a global rate coefficient of 5.9 x 10(-16) cm(3) molecule(-1) s(-1) was calculated, corresponding to the sum of geraniol-trans, 6-methyl-5-hepten-2-one, and 6-hydroxy-4-methyl-4-hexenal reactions with the ozone. These results are in good agreement with the experimental studies.