EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Studies on chemical constituents of Primula sikkmensis].
PMID 18589747
Abstract
To study the chemical constituents of Primula sikkmensis Hook. The chemical constituents of the plant were isolated and purified by column chromatography and their structures were elucidated by spectroscopic analysis. Seven compounds were isolated and identified as kaempferol, quercetin, Prunetin, hexacosanol, Apigenin, beta-sitosterol, quercetin-3-O-beta-D-glucoside, respectively. Compounds I--VII are obtained for the first time from this plant.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H2139
1-Hexacosanol, ≥97% (capillary GC)
C26H54O
82415
Prunetin, ≥98.0% (TLC)
C16H12O5