EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Chemical constituents of Changium smyrnioides].
PMID 18589748
Abstract
To study chemical constituents of Changium smyrnioides Wolff. The chemical components were isolated and purified by silica gel column and recrystallization. The chemical structures were elucidated on the basis of physico-chemical properties and spectral data. Ten compounds were isolated and identified as lignoceric acid (1), beta-sitosterol (2), stigmasterol (3), 5-hydroxy-8-methoxypsoralen (4), glycerylmonopalmitate (5), L-pyroglutamic acid (6), succinic acid (7), vanillic acid-4-O-beta-D-glucopyranoside (8 ), vanillic acid (9), daucosterol (10). Compounds 1, 4, 5, 6, 8 and 9 are obtained from the plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L6641
Lignoceric acid, ≥99% (capillary GC)
C24H48O2