EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis of quinolines from 3-formylchromone.
PMID 18593188
Abstract
A facile and versatile procedure for the synthesis of 3-(2-hydroxybenzoyl)quinolines and 7H-chromeno[3,2-c]quinolin-7-ones was elaborated on the basis of TMSCl-mediated recyclization of 3-formylchromone with various anilines. Limitations and scope of this methodology were established, and a possible mechanism for the heterocyclizations was proposed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
383449
3-Formylchromone, 97%
C10H6O3