EMAIL THIS PAGE TO A FRIEND
Spine
Association between the development of the body axis and the craniofacial skeleton studied by immunohistochemical analyses using collagen II, Pax9, Pax1, and Noggin antibodies.
PMID 18594453