EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Presence of flavin-containing monooxygenase in rat brain.
PMID 1859456
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
148229
Thiobenzamide, 98%
C7H7NS