EMAIL THIS PAGE TO A FRIEND
Journal of industrial microbiology & biotechnology
Production of a thermophilic maltooligosyl-trehalose synthase in Lactococcus lactis.
PMID 18594890
Abstract
The thermoacidophilic archaeon Sulfolobus solfataricus MT4 encodes a maltooligosyltrehalose synthase (MTS), that catalyzes an intramolecular transglycosylation process converting the glycosidic linkages at the reducing end of dextrins from alpha-1,4 into alpha-1,1. In this research the gene encoding MTS was cloned and expressed in Lactococcus lactis NZ9000 using the so-called NICE system. Growth conditions of the recombinant strain were optimized in flask experiments in relation to enzyme production. Batch experiments in 2 L-fermenters were performed on the best identified semidefined medium and 256 U L(-1) of recombinant MTS were produced. Purified recombinant MTS shows its optimal activity at 70 degrees C and pH 5.5, prefers maltoheptaose and maltohexaose as substrates, and demonstrates minimal side hydrolytic activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M9153
Maltohexaose, ≥65% (HPLC)
C36H62O31