EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Oxidative inactivation of protein tyrosine phosphatase 1B by organic hydroperoxides.
PMID 18595691
Abstract
Protein tyrosine phosphatases (PTPs) are cysteine-dependent enzymes that play a central role in cell signaling. Organic hydroperoxides cause thiol-reversible, oxidative inactivation of PTP1B in a manner that mirrors the endogenous signaling agent hydrogen peroxide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
269336
Peracetic acid solution, 32 wt. % in dilute acetic acid
C2H4O3