EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Subpicosecond transient signal spectroscopy of Prodan in dimethylformamide solution.
PMID 18596331
Abstract
We report a pump-probe experiment revealing the temporal evolution of subpicosecond evolution of Prodan's excited-state absorption in dimethylformamide. Also, we present calculation of the first spectral moment of this spectral band and estimation of different relaxation components on the subpicosecond time scale.
Related Materials