EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Hansch analysis of veratric acid derivatives as antimicrobial agents.
PMID 18597896
Abstract
The synthesis, characterization and antimicrobial evaluation of a new series of veratric acid derivatives are presented. Preliminary in vitro antimicrobial activity of the title compounds was assessed against a panel of microorganisms including Gram-positive and Gram-negative bacteria and fungi. Some of the veratric acid derivatives exhibited significant in vitro antimicrobial activity. QSAR investigation applied to find a correlation between different physicochemical parameters of the veratric acid derivatives and their antimicrobial activity indicated the importance of topological parameters in describing the antimicrobial activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D131806
3,4-Dimethoxybenzoic acid, ≥99%
C9H10O4