EMAIL THIS PAGE TO A FRIEND
Eukaryotic Cell
Aspergillus fumigatus RasA regulates asexual development and cell wall integrity.
PMID 18606827
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F3543
Fluorescent Brightener 28, used as a stain and brightening agent
C40H44N12O10S2