EMAIL THIS PAGE TO A FRIEND
The Journal of dermatological treatment
Perineal muco-cutaneous herpes zoster treated with brivudin.
PMID 18608718
Abstract
Herpes zoster results from the reactivation of latent varicella-zoster virus (VZV) from the dorsal root ganglion of sensory nerves. It is rarely described in the pediatric population. We report the case of an 8-year-old immunocompetent boy with a painful lumbosacral herpes zoster that was treated with brivudin and achieved rapid and sustained improvement in the absence of muco-cutaneous or pharmacological side effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B9647
(E)-5-(2-Bromovinyl)-2′-deoxyuridine
C11H13BrN2O5