EMAIL THIS PAGE TO A FRIEND
Thrombosis and haemostasis
Intoxication with three different superwarfarin compounds in an adult woman.
PMID 18612553
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32677
Difenacoum, PESTANAL®, analytical standard
C31H24O3