EMAIL THIS PAGE TO A FRIEND
Journal of neuroscience research
Functional contribution of alpha3L8' to the neuronal nicotinic alpha3 receptor.
PMID 18615639
Abstract
The role of position L8', located in transmembrane domain 1 of the neuronal nicotinic alpha3 subunit, was characterized by using two-electrode voltage clamp in Xenopus oocytes. Four amino acids (Ala, Ser, Phe, and Tyr) were inserted at this conserved position, and the mutant subunit was coexpressed with either wild-type beta2 or beta4 subunits. These substitutions led to significant alterations in the pharmacodynamic parameters of cholinergic agents, resulting in loss of function. Ala and Ser substitutions resulted in losses in agonist (ACh, nicotine, and DMPP) potency and intrinsic activity at both alpha3beta2 and alpha3beta4 receptors. Similarly, significant changes in antagonist potency were produced by the Ala and Ser substitutions. Phe and Tyr mutations did not alter the receptor's EC(50) for ACh or nicotine but reduced the EC(50) for DMPP at both receptors. The Phe mutation also reduced the intrinsic activity of all agonists tested at both receptors. The Tyr mutation, though, led to a decrease in intrinsic activity for all agonists at the alpha3beta2 receptor, yet resulted in no changes for DMPP, a decrease for nicotine, and an increase for ACh at the alpha3beta4 receptor. The most dramatic changes in the receptor's functional properties were produced by substitutions that introduced the largest changes in amino acid volume. Additional replacements (Gly, Thr, and Val) suggested an inverse correlation between amino acid volume at position alpha3L8' and EC(50) for alpha3beta4 nAChRs; however, alpha3beta2 nAChRs displayed a nonlinear correlation. These data demonstrate that structural alterations at position alpha3L8' could propagate to the agonist-binding site.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5891
1,1-Dimethyl-4-phenylpiperazinium iodide, ≥98% (TLC or titration), powder
C12H19IN2