EMAIL THIS PAGE TO A FRIEND
Organic letters
Expedient synthesis of pyrrolo[1,2-a]indoles: preparation of the core of yuremamine.
PMID 18616259
Abstract
Pyrrolo[1,2- a]indoles are conveniently prepared from tetrahydro-1,2-oxazines, which in turn are generated through the reaction of nitrones with 1,1-cyclopropanediesters. The synthetic route proves to be highly diastereoselective and provides access to the core of the recently discovered pyrrolo[1,2- a]indole natural product yuremamine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
343412
Cyclopropane-1,1-dicarboxylic acid, 97%
C5H6O4