EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Ortho-lithiation of free ferrocenyl alcohols: a new method for the synthesis of planar chiral ferrocene derivatives.
PMID 18622446
Abstract
An ortho-metalation method for free ferrocenyl alcohols has been developed, which allows preparation of planar chiral ferrocene derivatives with high yields and diastereoselectivities.