EMAIL THIS PAGE TO A FRIEND
Organic letters
Gold-catalyzed cycloisomerization of N-Propargylindole-2-carboxamides: application toward the synthesis of lavendamycin analogues.
PMID 18627172
Abstract
A series of N-propargylindole-2-carboxamides were found to undergo a AuCl 3-catalyzed cycloisomerization to give beta-carbolinones in high yield. The corresponding beta-chlorocarboline derivative was prepared and used for Pd(0)-catalyzed cross-coupling chemistry directed toward the synthesis of lavendamycin analogues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S1014
Streptonigrin from Streptomyces flocculus, ≥98%
C25H22N4O8