EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Ethyl 8-fluoro-6-(3-nitrophenyl)-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate as novel, highly potent, and safe antianxiety agent.
PMID 18637672
Abstract
Ethyl 8-fluoro-6-(4-nitrophenyl)- and ethyl 8-fluoro-6-(3-nitrophenyl)-4 H-imidazo[1,5-a][1,4]benzodiazepine 3-carboxylate 6 and 7 were synthesized as central benzodiazepine receptor (CBR) ligands and tested for their ability to displace [(3)H]flumazenil from bovine and human cortical brain membranes. Both compounds showed high affinity for bovine and human CBR. In particular, compound 7 emerged as the most interesting compound, having a partial agonist profile in vitro while possessing useful activity in various animal models of anxiety. In accordance with its partial agonist profile, compound 7 was devoid of typical benzodiazepine side effects. The homology model of the GABA A receptor developed by Cromer et al. was used to assess the binding modes of ligands 6 and 7. From our docking results, the partial agonist activity elicited by compound 7 is likely to be due to the 3'-nitro substituent, which is in the appropriate position to interact with Thr193 of the gamma 2-subunit by means of a hydrogen bond.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9261
Flunitrazepam
C16H12FN3O3