EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Fragrance material review on menthol racemic.
PMID 18640189
Abstract
A toxicologic and dermatologic review of menthol racemic when used as a fragrance ingredient is presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M0350000 Menthol, European Pharmacopoeia (EP) Reference Standard
C10H20O