EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Fragrance material review on 3,7-dimethyl-1-octanol.
PMID 18640235
Abstract
A toxicologic and dermatologic review of 3,7-dimethyl-1-octanol when used as a fragrance ingredient is presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W239100
3,7-Dimethyl-1-octanol, ≥98%, FCC, FG
C10H22O
305774
3,7-Dimethyl-1-octanol, ≥98%
C10H22O