EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Smqnr, a new chromosome-carried quinolone resistance gene in Stenotrophomonas maltophilia.
PMID 18644963
Abstract
A new chromosome-carried quinolone resistance gene from Stenotrophomonas maltophilia, Smqnr, was characterized. The gene was present in type strain CCUG 5866 and was also detected in 24 clinical isolates and showed some allelic diversity. The expression of Smqnr in Escherichia coli decreased the susceptibilities of the E. coli isolates to several fluoroquinolones.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56968
Sparfloxacin, ≥99.0% (HPLC)
C19H22F2N4O3