EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Observation of electrochemical single-electron-transfer events of gold nanoparticles in aqueous solution in the presence of both ammonium and sulfonate surface-active agents.
PMID 18646032
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
53272
Hexyltrimethylammonium bromide, ≥98.0% (AT)
C9H22BrN