EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Stereoselective synthesis of 3-alkyl-2-aryltetrahydrofuran-4-ols: total synthesis of (+/-)-paulownin.
PMID 18646863
Abstract
A formal [3 + 2]-cycloaddition involving the Lewis acid mediated reaction of alpha-silyloxy aldehydes and styrenes to afford 3-alkyl-2-aryltetrahydrofuran-4-ols has been developed. This methodology was applied to the total synthesis of the naturally occurring furofuran lignan (+/-)-paulownin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00206
Paulownin, ≥95% (LC/MS-ELSD)
C20H18O7