EMAIL THIS PAGE TO A FRIEND
Chemical Society reviews
A new trend in rhodamine-based chemosensors: application of spirolactam ring-opening to sensing ions.
PMID 18648672
Abstract
This tutorial review focuses on the recent development of rhodamine derivatives, in which the spirolactam (non-fluorescent) to ring-opened amide (fluorescent) process was utilized.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
59746 Atto Rho14