EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
Guiding the self-assembly of a second-generation polyphenylene dendrimer into well-defined patterns.
PMID 18651629
Abstract
A second-generation polyphenylene dendrimer 1 is shown to self-assemble into nanofibers. To guide the formation of the dendrimer fibers into well-defined patterns, 1H,1H,2H,2H-perfluorodecyltrichlorosilane is grafted in the gas phase onto a silicon substrate. De-wetting of the solution on the nanopatterned surface results in the formation of a nanostructured template, into which fiber growth subsequently occurs under the constraints set by the de-wetted morphology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
175552
Trichlorosilane, 99%
HCl3Si