EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Controlled cyclopolymerization through quantitative 19-membered ring formation.
PMID 18652466
Abstract
Cyclopolymerization of a bis-methacrylate monomer derived from trans-cyclohexanediol and 2-methacryloyloxyethyl isocyanate proceeded through 19-membered ring formation. The unusual large ring formation originated from the designed steric regulation by the cyclohexane ring and the hydrogen bonds. The RAFT polymerization employing cumyl dithiobenzoate attained controlled polymerization, which supported the quantitative cyclization.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
477060
2-Isocyanatoethyl methacrylate, contains ≤0.1% BHT as inhibitor, 98%
C7H9NO3