EMAIL THIS PAGE TO A FRIEND
Langmuir : the ACS journal of surfaces and colloids
Modification of carbon electrode with aryl groups having an aliphatic amine by electrochemical reduction of in situ generated diazonium cations.
PMID 18652500
Abstract
The electrochemically induced functionalization of glassy carbon electrode by aryl groups having an aliphatic amine group was achieved by reduction of in situ generated diazonium cations in aqueous media. The corresponding diazonium cations of 4-aminobenzylamine, 2-aminobenzylamine, 4-(2-aminoethyl)aniline, N-methyl-1,2-phenylenediamine, and N, N-dimethyl- p-phenylenediamine were generated in situ with sodium nitrite in aqueous HCl. The kinetics of electrochemical grafting were investigated with electrochemical impedance spectroscopy and electrochemical quartz crystal microbalance measurements (with carbon-coated quartz crystal), and the barrier properties of the grafted layers were evaluated by cyclic voltammetry in the presence of electroactive redox probes such as Fe(CN)6 3-/4- and Ru(NH 3)6 (3+). The grafting efficiency of aryl groups was found to depend on the nature of the amine (primary, secondary, and tertiary), the chain length of the alkyl substituent, and the substitution position on the aromatic ring. The nitrosation of the "aliphatic" amine, in the case of secondary and tertiary amines, was also evidenced by X-ray photoelectron spectroscopy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
368466
4-Aminobenzylamine, 99%
C7H10N2
398985
N-Methyl-1,2-phenylenediamine, 97%
C7H10N2