EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Total synthesis of malformin C, an inhibitor of bleomycin-induced G2 arrest.
PMID 18653995
Abstract
Total synthesis of a fungal cyclic peptide, malformin C, recently rediscovered as a G2 checkpoint inhibitor was completed. Our synthesis involved a convergent approach with respect to a linear pentapeptide, cyclization, and oxidative disulfide formation.