EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Reversible modulation in luminescence intensity of a single vesicle composed of diblock azo-copolymer and tris(dibenzoylmethanate)(phenanthroline)europium(III).
PMID 18656420
Abstract
Tris(dibenzoylmethanate)(phenanthroline)europium(III)[Eu(DBM)(3)Phen]-doped amphiphilic vesicles were obtained by self-assembling of poly(N-isopropylacrylamide)-b-poly[6-[4-(4-methylphenyl-azo) phenoxy] hexylacrylate] (PNIPAM(83)-b-PAzoM(20)) in presence of Eu(DBM)(3)Phen in the mixed solvent of THF/H(2)O (50/50 vol.%). Their optical properties were studied by UV-vis and fluorescence spectroscopies. The UV-vis spectrum showed that the electronic transition bands of azobenzene and Eu(DBM)(3)Phen were overlapped at about 365 nm and the main peak of fluorescence emission band appeared at 612 nm. So the vesicles showed obvious red luminescence. It was found that the fluorescence intensity of a single Eu(DBM)(3)Phen-doped vesicle could be modulated by irradiation with UV and visible light due to the reversible trans-cis-trans photoisomerization reaction of azobenzene moiety. Possible energy allocation process for this property was discussed in details.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D33454
1,3-Diphenyl-1,3-propanedione, 98%
C15H12O2