EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Solid-liquid transfer of biophenols from olive leaves for the enrichment of edible oils by a dynamic ultrasound-assisted approach.
PMID 18656923
Abstract
A continuous approach assisted by ultrasound for direct enrichment of edible oils (olive, sunflower, and soya) with the main phenols in olive leaves (i.e., oleuropein, verbascoside, apigenin-7-glucoside, and luteolin-7-glucoside) has been developed. Multivariate methodology was used to carry out a detailed optimization of the enrichment, and quantitation of the transferred compounds was based on LC-MS-MS in multiple reaction monitoring optimizing the most sensitive transition for each biophenol. Under the optimal working conditions, only 20 min is necessary to enrich the edible oils with 14.45-9.92 microg/mL oleuropein, 2.29-2.12 microg/mL verbascoside, 1.91-1.51 microg/mL apigenin-7-glucoside, and 1.60-1.42 microg/mL luteolin-7-glucoside. The enrichment method is carried out at room temperature and is organic-solvent-free; thus, the healthy properties of the edible oils improve as does their quality. Also, the low acquisition and maintenance costs of an ultrasound source and its application in a dynamic system make advisable the industrial implementation of the proposed method.