EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Purification, molecular cloning, and enzymatic properties of a family 12 endoglucanase (EG-II) from fomitopsis palustris: role of EG-II in larch holocellulose hydrolysis.
PMID 18658283
Abstract
A family 12 endoglucanase with a molecular mass of 23,926 Da (EG-II) from the brown-rot basidiomycete Fomitopsis palustris was purified and characterized. One of the roles of EG-II in wood degradation is thought to be to loosen the polysaccharide network in cell walls by disentangling hemicelluloses that are associated with cellulose.
Related Materials