EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
High-performance liquid chromatography assays for desmethoxyyangonin, methysticin, kavain and their microsomal metabolites.
PMID 18661482
Abstract
Three novel, simple and reproducible high-performance liquid chromatography quantitative assays with UV detection were developed and validated for three major kavalactones--desmethoxyyangonin, methysticin and kavain--in rat liver microsomes using diazepam as an internal standard; liquid-liquid extraction was used for sample preparation and analysis was performed on a Shimadzu 10A high-performance liquid chromatography system. The analysis was carried out in reversed-phase mode with a Luna C(18) column (150 x 2.00 mm, 3 microm) at 40 degrees C. The limit of quantitation was 0.1 microg/mL using 0.25 mL of microsomal solution. The assays were linear over the range 0.1-10 microg/mL for desmethoxyyangonin, methysticin and kavain. Quality control samples exhibited good accuracy and precision with relative standard deviations lower than 15% and recoveries between 85 and 105%. The assays exhibited satisfactory performance with high sensitivity for quantifying desmethoxyyangonin, methysticin and kavain in rat liver microsomes and were successfully used to determine the three kavalactones and their microsomal metabolites.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
59780
DL-Kavain, analytical standard
C14H14O3
80488
R-(+)-Methysticin, analytical standard
C15H14O5