EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Iterative molecular assembly based on the cation-pool method. Convergent synthesis of dendritic molecules.
PMID 18661982
Abstract
An iterative method for molecular assembly has been developed based on the cation-pool method using (trimethylsilyl)diphenylmethane as a building block. The silyl group works as both an activating group of the benzene ring in the Friedel-Crafts type reaction and an electroauxiliary in the subsequent low temperature anodic oxidation to generate dendritic diarylcarbenium ions, which were well characterized by low-temperature NMR spectroscopy. The convergent synthesis of dendritic molecules has been achieved by repeating the sequence.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D209317
Diphenylmethane, 99%
C13H12