EMAIL THIS PAGE TO A FRIEND
Przeglad lekarski
[A case of Salmonella paratyphi A infection in Poland].
PMID 18663912
Abstract
Paratyphoid fever is an acute infection caused by Salmonella paratyphi A, B or C. The disease is transmitted from person to person by fecal-oral way. Typical for typhoid fever are splenomegaly, bradycardia, fever, constipation or mild diarrhoea oftten associated with abdominal tenderness. We present the case of patient who was infected by Salmonella paratyphi C while his travelling in Asia.
Related Materials