EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Analysis of captan, folpet, and captafol in apples by dispersive liquid-liquid microextraction combined with gas chromatography.
PMID 18665351
Abstract
A novel method was developed for the determination of captan, folpet, and captafol in apples by dispersive liquid-liquid microextraction (DLLME) coupled with gas chromatography-electron capture detection (GC-ECD). Some experimental parameters that influence the extraction efficiency, such as the type and volume of the disperser solvents and extraction solvents, extraction time, and addition of salt, were studied and optimized to obtain the best extraction results. Under the optimum conditions, high enrichment factors for the compounds were achieved ranging from 824 to 912. The recoveries of fungicides in apples at spiking levels of 20.0 microg kg(-1) and 70.0 microg kg(-1) were 93.0-109.5% and 95.4-107.7%, respectively. The relative standard deviations (RSDs) for the apple samples at 30.0 microg kg(-1) of each fungicide were in the range from 3.8 to 4.9%. The limits of detection were between 3.0 and 8.0 microg kg(-1). The linearity of the method ranged from 10 to 100 microg kg(-1) for the three fungicides, with correlation coefficients (r (2)) varying from 0.9982 to 0.9997. The obtained results show that the DLLME combined with GC-ECD can satisfy the requirements for the determination of fungicides in apple samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45365
Captafol, PESTANAL®, analytical standard
C10H9Cl4NO2S
32057
Folpet, PESTANAL®, analytical standard
C9H4Cl3NO2S